missing translation for 'onlineSavingsMsg'
Learn More

Sodium iodide hydrate, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals

£42.11 - £329.64

Chemical Identifiers

MDL Number MFCD00003532
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11367027
SDS View Documents
Thermo Scientific Alfa Aesar
010871.06
5 g
£42.11
5g
Please to purchase this item. Need a web account? Register with us today!
11387027
SDS View Documents
Thermo Scientific Alfa Aesar
010871.14
25 g
£111.56
25g
Please to purchase this item. Need a web account? Register with us today!
11377027
SDS View Documents
Thermo Scientific Alfa Aesar
010871.22
100 g
£329.64
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Sodium Iodide is used in the synthesis of dye-sensitized solar cells. Also used in the preparation of propargylamines as inhibitors of lysine specific. Demethylase. Used in photography and as analytical reagents. They are used in the measurement of the energy of gamma rays, by measuring the amplitude of pulses of light generated by electrons which are excited by the gamma rays and in the Finkelstein reaction via an SN2 pathway, detection of radiation, and in the conversion of an alkyl chloride into an alkyl iodide. Sodium Iodide is used in PCR and to produce chemokines, particularly CCL2, CXCL8, and CXC14 by stimulation of thyroid follicular cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

MFCD00003532

Specifications

Crystalline Aggregates
Soluble in water (1840mg/ml at 25°C),alcohol,glycerol,and acetone. Insoluble in ether.
99.999%
Hygroscopic
Sodium iodide hydrate
MFCD00003532
149.89 (Anhydrous)
Puratronic™
(metals basis)
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362

H315-H319

missing translation for 'dotInformation' : Hazard Class: 9; Packaging Group: III

missing translation for 'einecsNumber' : 231-679-3

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.