missing translation for 'onlineSavingsMsg'
Learn More

Sodium hexanoate, 99%, Thermo Scientific Chemicals

£61.10 - £120.00

Chemical Identifiers

CAS 10051-44-2
Molecular Formula C6H11NaO2
Molecular Weight (g/mol) 138.14
MDL Number MFCD00059056
InChI Key UDWXLZLRRVQONG-UHFFFAOYSA-M
Synonym sodium hexanoate, sodium caproate, hexanoic acid, sodium salt, hexanoic acid sodium salt, sodium capronate, sodium n-caproate, sodium n-hexanoate, unii-e00cg11s66, caproic acid sodium salt, hexanoic acid, sodium salt 1:1
PubChem CID 4087444
IUPAC Name sodium;hexanoate
SMILES [Na+].CCCCCC([O-])=O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10585100
View Documents
Thermo Scientific Acros
437540100
10 g
£61.10
10g
Please to purchase this item. Need a web account? Register with us today!
10038289
View Documents
Thermo Scientific Acros
437540250
25 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

10051-44-2
138.14
UDWXLZLRRVQONG-UHFFFAOYSA-M
4087444
[Na+].CCCCCC([O-])=O
C6H11NaO2
MFCD00059056
sodium hexanoate, sodium caproate, hexanoic acid, sodium salt, hexanoic acid sodium salt, sodium capronate, sodium n-caproate, sodium n-hexanoate, unii-e00cg11s66, caproic acid sodium salt, hexanoic acid, sodium salt 1:1
sodium;hexanoate

Specifications

231.2°C to 233.8°C
White
99%
MFCD00059056
UDWXLZLRRVQONG-UHFFFAOYSA-M
sodium;hexanoate
4087444
99%
2% max. ()
Sodium hexanoate
10051-44-2
Powder
C6H11NaO2
sodium hexanoate, sodium caproate, hexanoic acid, sodium salt, hexanoic acid sodium salt, sodium capronate, sodium n-caproate, sodium n-hexanoate, unii-e00cg11s66, caproic acid sodium salt, hexanoic acid, sodium salt 1:1
[Na+].CCCCCC([O-])=O
138.14
138.14
Authentic
Glass Bottle
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Warning

missing translation for 'einecsNumber' : 233-179-

Product Identifier
  • Sodium hexanoate, 99%

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only