missing translation for 'onlineSavingsMsg'
Learn More

Sodium hexachloroplatinate(IV) hexahydrate, Thermo Scientific Chemicals

For Use As a Catalyst

£76.68 - £293.46

Chemical Identifiers

CAS 19583-77-8
Molecular Formula Cl6H12Na2O6Pt
Molecular Weight (g/mol) 561.85
MDL Number MFCD00149172
InChI Key XGFSWVNCHVSCSP-UHFFFAOYSA-H
Synonym sodium hexachloroplatinate iv hexahydrate, cl6pt.2na.6h2o, platinum iv sodium chloride, disodium;hexachloroplatinum 2-;hexahydrate, disodium hexachloroplatinate 2-hexahydrate
PubChem CID 24846071
SMILES O.O.O.O.O.O.[Na+].[Na+].Cl[Pt](Cl)(Cl)(Cl)(Cl)Cl
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11361688
View Documents
Thermo Scientific Alfa Aesar
011047.03
1 g
£76.68
1g
Please to purchase this item. Need a web account? Register with us today!
11371688
View Documents
Thermo Scientific Alfa Aesar
011047.06
5 g
£293.46
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Catalyst.Sodium hexachloroplatinate(IV) hexahydrate is used as a catalyst. It acts as a source of platinum in the preparation of platinum complex. It is also used in the synthesis of platinum nanoparticles using glutathione as a reducing agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

19583-77-8
561.85
XGFSWVNCHVSCSP-UHFFFAOYSA-H
24846071
Cl6H12Na2O6Pt
MFCD00149172
sodium hexachloroplatinate iv hexahydrate, cl6pt.2na.6h2o, platinum iv sodium chloride, disodium;hexachloroplatinum 2-;hexahydrate, disodium hexachloroplatinate 2-hexahydrate
O.O.O.O.O.O.[Na+].[Na+].Cl[Pt](Cl)(Cl)(Cl)(Cl)Cl

Specifications

110°C-6H2O
Orange
Cl6H12Na2O6Pt
sodium hexachloroplatinate iv hexahydrate, cl6pt.2na.6h2o, platinum iv sodium chloride, disodium;hexachloroplatinum 2-;hexahydrate, disodium hexachloroplatinate 2-hexahydrate
XGFSWVNCHVSCSP-UHFFFAOYSA-H
disodium;hexachloroplatinum(2-);hexahydrate
24846071
Odorless
2.5
19583-77-8
Crystalline
MFCD00149172
Soluble in water,alcohol. Insoluble in ether.
O.O.O.O.O.O.[Na+].[Na+].Cl[Pt](Cl)(Cl)(Cl)(Cl)Cl
561.85
561.88 (453.79 Anhydrous)
Orange powder
Sodium hexachloroplatinate(IV) hexahydrate
Videos
Safety and Handling

Safety and Handling

P261-P272-P280g-P285-P302+P352-P304+P341-P333+P313-P342+P311-P363-P501c

H317-H334

missing translation for 'dotInformation' : DOT Class: 6.1, Packaging Group: III

missing translation for 'einecsNumber' : 240-983-5

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

Product Identifier
  • Sodium hexachloroplatinate(IV) hexahydrate

Signal Word
  • Danger

Hazard Category
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1

Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.

Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : Sodium hexachloroplatinate(IV) hexahydrate

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only