missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Dodecylsulfate, 10% Solution, Ultrapure, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 mL
500 mL
1 L
Packaging:
Nalgene bottle
Specifications
Specifications
Physical Form | Solution |
Concentration | 10% Solution |
Quantity | 500 mL |
Packaging | Nalgene bottle |
IUPAC Name | sodium dodecyl sulfate |
Chemical Name or Material | Sodium Dodecylsulfate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'