missing translation for 'onlineSavingsMsg'
Learn More
Sodium Dichromate Dihydrate, puriss. p.a. ACS Reagent, ≥99.5% (RT), Honeywell Fluka™ Product Code.: 15610520

Sodium Dichromate Dihydrate, puriss. p.a. ACS Reagent, ≥99.5% (RT), Honeywell Fluka™

Product Code. 15610520 Shop All Honeywell Fluka Products
1 kg, Poly Bottle
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Poly Bottle
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Product Code. 15610520

Brand: Honeywell-Fluka 006241KG

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Puriss. p.a., ACS reagent,≥99.5% (RT)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7789-12-0
Molecular Formula Cr2H4Na2O9
Molecular Weight (g/mol) 297.995
MDL Number MFCD00149166
InChI Key JYDRNIYTFCBIFC-UHFFFAOYSA-N
Synonym sodium dichromate dihydrate, unii-52125xyy2a, ccris 6344, disodium chromate dihydrate, sodium dichromate vi dihydrate, sodium dichromate dihydrate vi, chromic acid h2cr2o7 , disodium salt, dihydrate, dichromic acid h2cr2o7 , disodium salt, dihydrate, dsstox_cid_12061, dsstox_rid_78903Show MoreShow Less
PubChem CID 108005
IUPAC Name disodium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium;dihydrate
SMILES O.O.[O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-].[Na+].[Na+]

Specifications

Melting Point 91°C (literature)
Physical Form Solid
Quantity 1 kg
Linear Formula Na2Cr2O7 · 2H2O
UN Number UN3086
Formula Weight 298g/mol
Percent Purity ≥99.5% (RT)
Grade ACS/puriss. p.a.
Packaging Poly Bottle
Density 2.350g/cu.cm
Chemical Name or Material Sodium Dichromate Dihydrate
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sodium Dichromate Dihydrate, puriss. p.a. ACS Reagent, ≥99.5% (RT), Honeywell Fluka™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.