missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STE Buffer, Sodium Chloride-Tris-EDTA, 10X Solution, pH 8.0, Fisher BioReagents
Brand: Fisher Bioreagents BP2479-1
Code : 32
Additional Details : Weight : 1.05000kg
Description
- Save time and simplify your buffer preparation step by using Fisher BioReagents 10X STE buffer solution - simply dilute as needed
- When diluted, 1X solution contains 10mM tris, 1mM EDTA, and 100mM NaCl with pH 8.0 at 25°C
- Filtered through a 5 micron filter and autoclaved
- Tested for the absence of DNase, RNase, and protease
- STE buffer is used for DNA extraction protocols
- Intended for laboratory research use only
Specifications
Sodium Chloride-Tris-EDTA (STE) Buffer | |
7647-14-5,60-00-4,7647-01-0,7732-18-5 | |
Undesignated | |
Liquid | |
DNase free | |
Water:>90% Sodium Chloride: <10% 1,3-Propanediol, 2-amino-2-(hydroxymethyl)-hydrochloride: <5% Ethylenediamine tetraacetic acid (EDTA): <1.0% | |
Protease free |
10X, pH 8.0 | |
100°C | |
8 | |
1 L | |
Filtered through a 5-micron filter. | |
Nalgene Poly Bottle | |
Saline Tris–EDTA, STE, TEN |