missing translation for 'onlineSavingsMsg'
Learn More
Sodium chloride, Puriss., meets analytical spec. of BP, Ph. Eur., USP, 99.0-100.5% (calc. to the dried substance), ≤0.00002% Al, Honeywell Fluka™ Product Code.: 15691300

Sodium chloride, Puriss., meets analytical spec. of BP, Ph. Eur., USP, 99.0-100.5% (calc. to the dried substance), ≤0.00002% Al, Honeywell Fluka™
BIOPHARMA

Product Code. 15691300 Shop All Honeywell Fluka Products
25 kg
Click to view available options
Quantity:
1 kg
5 kg
10 kg
25 kg
32 kg
4 x 5 kg
6 x 1 kg
This item is not returnable. View return policy

Product Code. 15691300

Brand: Honeywell-Fluka 1342325KG

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Puriss., meets analytical specification of BP, Ph. Eur., USP, 99.0-100.5% (calc. to the dried substance), ≤0.00002% Al

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7647-14-5
Molecular Formula ClNa
Molecular Weight (g/mol) 58.44
MDL Number MFCD00003477
InChI Key FAPWRFPIFSIZLT-UHFFFAOYSA-M
Synonym sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex
PubChem CID 5234
ChEBI CHEBI:26710
SMILES [Na+].[Cl-]

Specifications

Boiling Point 1,465°C
Melting Point 801°C (literature)
CAS Max % ≤100.0000%
Quantity 25 kg
Assay Percent Range 99.0 to 100.5%
Linear Formula NaCl
UN Number NONH for all modes of transport
Beilstein 3534976
IUPAC Name sodium chloride
Formula Weight 58.44
Percent Purity 99.0 to 100.5% (calc. to the dried substance)
Grade puriss.
Packaging Paper / Poly Bag
Density 2.160 g/cm3
Chemical Name or Material Sodium chloride, Puriss., meets analytical specification of BP, Ph. Eur., USP, 99.0-100.5% (calc. to the dried substance), ≤0.00002% Al
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sodium chloride, Puriss., meets analytical spec. of BP, Ph. Eur., USP, 99.0-100.5% (calc. to the dried substance), ≤0.00002% Al, Honeywell Fluka™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.