Learn More
Sodium chlorate, ACS, 99.0% min, Thermo Scientific™
Maximum level of impurities: Insoluble matter 0.005%, BrO3 0.015%, Cl 0.005%, SO4 0.001%, Heavy metals (as Pb) 0.001%, Fe 5ppm, Ca 0.002%, Mg 0.002%, K 0.01%
£29.25 - £243.21
Chemical Identifiers
CAS | 7775-09-9 |
---|---|
Molecular Formula | ClNaO3 |
Molecular Weight (g/mol) | 106.44 |
MDL Number | MFCD00003479 |
InChI Key | YZHUMGUJCQRKBT-UHFFFAOYSA-M |
Synonym | sodium chlorate, chloric acid, sodium salt, asex, chlorate de sodium, chlorsaure, tumbleleaf, atlacide, desolet, kusatol, rasikal |
PubChem CID | 516902 |
ChEBI | CHEBI:65242 |
IUPAC Name | sodium;chlorate |
SMILES | [O-]Cl(=O)=O.[Na+] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11448600
|
Thermo Scientific Alfa Aesar
044408.22 |
100g |
£29.25
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11458600
|
Thermo Scientific Alfa Aesar
044408.36 |
500g |
£72.76
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11468600
|
Thermo Scientific Alfa Aesar
044408.A3 |
£243.21
2kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
7775-09-9 | |
106.44 | |
YZHUMGUJCQRKBT-UHFFFAOYSA-M | |
516902 | |
sodium;chlorate |
ClNaO3 | |
MFCD00003479 | |
sodium chlorate, chloric acid, sodium salt, asex, chlorate de sodium, chlorsaure, tumbleleaf, atlacide, desolet, kusatol, rasikal | |
CHEBI:65242 | |
[O-]Cl(=O)=O.[Na+] |
Specifications
7775-09-9 | |
ClNaO3 | |
sodium chlorate, chloric acid, sodium salt, asex, chlorate de sodium, chlorsaure, tumbleleaf, atlacide, desolet, kusatol, rasikal | |
YZHUMGUJCQRKBT-UHFFFAOYSA-M | |
sodium;chlorate | |
516902 | |
106.45 | |
Odorless | |
Sodium chlorate, For analysis ACS |
99% min. | |
MFCD00003479 | |
Soluble in water,glycerol,hydrazine,acetone and methanol. Slightly soluble in ethanol and ammonia. | |
[O-]Cl(=O)=O.[Na+] | |
106.44 | |
CHEBI:65242 | |
ACS | |
2.5 g/cm3 at 20°C |
Safety and Handling
P210-P220-P221-P260-P264b-P270-P280-P283-P301+P312-P306+P360-P314-P330-P370+P378q-P371+P380+P375-P501c
H271-H302-H373
missing translation for 'dotInformation' : DOT Class: 5.1, Packaging Group: II
missing translation for 'einecsNumber' : 231-887-4
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Sodium chlorate
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 2
- Oxidising solid Category 1
Hazard Statement
- H271-May cause fire or explosion; strong oxidiser.
- H302-Harmful if swallowed.
- H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P220-Keep/Store away from clothing/combustible materials.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P371+P380+P375-In case of major fire and large quantities: Evacuate area Fight fire remotely due to the risk of explosion.
Supplemental information
- MIXTURE LIST-Contains : Sodium chlorate
RUO – Research Use Only