missing translation for 'onlineSavingsMsg'
Learn More
Sodium Carbonate Decahydrate, Extra Pure, SLR, Fisher Chemical™ Product Code.: 10785531

Sodium Carbonate Decahydrate, Extra Pure, SLR, Fisher Chemical™

Product Code. 10785531
1 kg, Plastic powder jar
Click to view available options
Quantity:
1 kg
2.5 kg
5 kg
Packaging:
Plastic keg
Plastic powder jar
This item is not returnable. View return policy

Product Code. 10785531

Brand: Fisher Chemical S/2720/60

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 6132-02-1
Molecular Formula CH20Na2O13
Molecular Weight (g/mol) 286.14
MDL Number 149178
InChI Key XYQRXRFVKUPBQN-UHFFFAOYSA-L
Synonym sodium carbonate decahydrate, natrii carbonas decahydricus, carbonic acid disodium salt, decahydrate, unii-ls505bg22i, disodium decahydrate carbonate, acmc-1b9jt, natrium-carbonat-dekahydrat, natrium-carbonat-10-wasser, disodium carbonate decahydrate, ksc353a3t
PubChem CID 151402
IUPAC Name disodium;carbonate;decahydrate
SMILES C(=O)([O-])[O-].O.O.O.O.O.O.O.O.O.O.[Na+].[Na+]

Specifications

Melting Point 34°C
Color White
pH 11 to 12
Physical Form Solid
Quantity 1 kg
Formula Weight 286.01g/mol
Packaging Plastic powder jar
compliance-icons
Product Identifier
  • Sodium carbonate decahydrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : Sodium carbonate decahydrate, Sodium carbonate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sodium Carbonate Decahydrate, Extra Pure, SLR, Fisher Chemical™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.