missing translation for 'onlineSavingsMsg'
Learn More

Sodium Benzoate, N.F. - F.C.C., J.T. Baker™

Chemical Identifiers

CAS 532-32-1
Molecular Formula C7H5NaO2
Molecular Weight (g/mol) 144.11
MDL Number MFCD00012463
InChI Key WXMKPNITSTVMEF-UHFFFAOYSA-M
Synonym sodium benzoate, benzoic acid, sodium salt, benzoic acid sodium salt, sobenate, antimol, benzoate sodium, benzoate of soda, benzoate, sodium, natrium benzoicum, caswell no. 746
PubChem CID 517055
IUPAC Name sodium benzoate
SMILES [Na+].[O-]C(=O)C1=CC=CC=C1
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
15538134
View Documents
Avantor J.T.Baker
3500-01
500 g Wide Mouth Amber Glass Bottle N/A
Please to purchase this item. Need a web account? Register with us today!
15548134
View Documents
Avantor J.T.Baker
3500-05
2.5 kg Poly Bottle N/A
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

532-32-1
144.11
WXMKPNITSTVMEF-UHFFFAOYSA-M
517055
[Na+].[O-]C(=O)C1=CC=CC=C1
C7H5NaO2
MFCD00012463
sodium benzoate, benzoic acid, sodium salt, benzoic acid sodium salt, sobenate, antimol, benzoate sodium, benzoate of soda, benzoate, sodium, natrium benzoicum, caswell no. 746
sodium benzoate

Specifications

532-32-1
MFCD00012463
WXMKPNITSTVMEF-UHFFFAOYSA-M
sodium benzoate
517055
FCC/NF
Sodium Benzoate
C7H5NaO2
sodium benzoate, benzoic acid, sodium salt, benzoic acid sodium salt, sobenate, antimol, benzoate sodium, benzoate of soda, benzoate, sodium, natrium benzoicum, caswell no. 746
[Na+].[O-]C(=O)C1=CC=CC=C1
144.11
144.11
1L = 1.44kg
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.