Learn More
Thermo Scientific Chemicals Sodium azide, 99+%, for biochemistry
Sodium azide, CAS # 26628-22-8, is the sodium salt of hydrogen azide (hydrazoic acid). This compound shows antibacterial and mutagenic activities. It also can inhibit the mitochondrial respiratory chain.
£30.85 - £316.58
Chemical Identifiers
CAS | 26628-22-8 |
---|---|
Molecular Formula | N3Na |
Molecular Weight (g/mol) | 65.01 |
InChI Key | PXIPVTKHYLBLMZ-UHFFFAOYSA-N |
Synonym | sodium azide, azide, sodium, natriumazid, nemazyd, smite, kazoe, azoture de sodium, sodium azide na n3, hydrazoic acid, sodium salt, natriumazid german |
PubChem CID | 33557 |
ChEBI | CHEBI:278547 |
IUPAC Name | sodium;azide |
SMILES | [N-]=[N+]=[N-].[Na+] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
12695107
|
Thermo Scientific Chemicals
447810250 |
25 g |
£30.85
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
12605117
|
Thermo Scientific Chemicals
447811000 |
100 g |
£86.83
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
12615117
|
Thermo Scientific Chemicals
447815000 |
500 g |
£316.58
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Information
• Sodium azide is a precursor for preparing inorganic azides such as lead azide and silver azide
• One mechanism of action associated with this compound can be related to the inhibition of mitochondrial ATPase, that consequently suppresses oxidative phosphorylation
Applications
• Sodium azide is an inorganic compound with antibacterial and mutagenic activities mainly used in biological samples
• Sodium azide inhibits cytochrome oxidase in gram-negative bacteria. However, its antibacterial capacity is more limited in some gram-positive bacteria (streptococci, pneumococci, lactobacilli), presenting intrinsic resistance
• Sodium azide is used for the introduction of an azide group to prepare a desired compound
• It is useful in biochemical approaches as a probe reagent and a preservative
• This compound can be used to synthetize blue fluorescent copolymers and metal phosphonates
Chemical Identifiers
26628-22-8 | |
65.01 | |
sodium azide, azide, sodium, natriumazid, nemazyd, smite, kazoe, azoture de sodium, sodium azide na n3, hydrazoic acid, sodium salt, natriumazid german | |
CHEBI:278547 | |
[N-]=[N+]=[N-].[Na+] |
N3Na | |
PXIPVTKHYLBLMZ-UHFFFAOYSA-N | |
33557 | |
sodium;azide |
Specifications
300.0°C | |
26628-22-8 | |
N3Na | |
15,8717 | |
Solubility in water: 420g/L (17°C). Other solubilities: soluble in liquid ammonia, slightly soluble in alcohol, insoluble in diethyl ether | |
[N-]=[N+]=[N-].[Na+] | |
65.01 | |
CHEBI:278547 | |
≥99% | |
Plastic bottle | |
1.8500g/mL |
275.0°C | |
99% min. (Argentometry) | |
NaN3 | |
sodium azide, azide, sodium, natriumazid, nemazyd, smite, kazoe, azoture de sodium, sodium azide na n3, hydrazoic acid, sodium salt, natriumazid german | |
PXIPVTKHYLBLMZ-UHFFFAOYSA-N | |
sodium;azide | |
33557 | |
65.01 | |
Biochemical | |
1.85 | |
Sodium azide |
Safety and Handling
GHS H Statement
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Very toxic to aquatic life with long lasting effects.
GHS P Statement
Rinse mouth.
Wear protective gloves/protective clothing.
IF ON SKIN: Gently wash with plenty of soap and water.
Immediately call a POISON CENTER or doctor/physician.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 247-852-1
- Sodium azide
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 2
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
Hazard Statement
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- H300-Fatal if swallowed.
- EUH032-Contact with acids liberates very toxic gas.
Precautionary Statement
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/.
RUO – Research Use Only