missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SMU1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15511-1-AP-20UL
This item is not returnable.
View return policy
Description
This antibody is a rabbit polyclonal antibody raised against a region of human REDD1 antigen.
SMU1 (suppressor of Mec8 and Unc-52 homolog), also known as BWD (brain-enriched WD), is a member of the WD-repeat SMU1 family and contains one CTLH domain, one LisH domain and six WD-repeats. SMU1 is a homolog of the C. elegans protein Smu1, a ubiquitously expressed nuclear protein that is believed to play a role in alternative splicing events of Unc-52. In mammals, SMU1 is a component of the spliceosome and appears to exhibit a conserved function, affecting the splicing of the mammalian Unc-52 homolog, namely Perlecan. Mutations in the gene encoding SMU1 may affect the function of the spliceosome.Specifications
SMU1 | |
Polyclonal | |
Unconjugated | |
SMU1 | |
BWD, fSAP57, RP11 54K16.3, SMU 1, SMU1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
117541, 55234, 74255 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry, Western Blot, Immunofluorescence | |
0.21 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q2TAY7, Q3UKJ7, Q99M63 | |
SMU1 | |
SMU1 Fusion Protein Ag7688 | |
20 μL | |
Primary | |
Mouse, Rat, Human | |
Antibody | |
IgG |