missing translation for 'onlineSavingsMsg'
Learn More

SMPD1/ASM Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

£135.00 - £349.00

Specifications

Antigen SMPD1/ASM
Concentration 0.14 mg/mL
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16823334
View Documents
Proteintech
14609-1-AP-20UL
20 μL
£135.00
20µL
Please to purchase this item. Need a web account? Register with us today!
16813334
View Documents
Proteintech
14609-1-AP-150UL
150 μL
£349.00
150µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The protein encoded by this gene is a lysosomal acid sphingomyelinase that converts sphingomyelin to ceramide. The encoded protein also has phospholipase C activity. Defects in this gene are a cause of Niemann-Pick disease type A (NPA) and Niemann-Pick disease type B (NPB). Three transcript variants encoding two different isoforms have been found for this gene.
Specifications

Specifications

SMPD1/ASM
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
Rabbit
Mouse, Rat, Human
P17405, Q04519
20597, 308909, 6609
SMPD1,ASM Fusion Protein Ag6180
Primary
-20°C
SMPD1
0.14 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
ASM, SMPD1
SMPD1
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications