missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SMO Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20787-1-AP-20UL
This item is not returnable.
View return policy
Description
Smoothened drosophila homolog (SMOH) is a serpentine protein belonging to the G-protein coupled receptor Fz/Smo family with 787 aa, seven transmembrane G protein-coupled receptors with 4 glycosylation sites and a putative extracellular amino terminus. Smoothened activated by b-Arrestin 2 and GRK2 acts as a receptor for Shh, the Sonic hedgehog gene product, and is a positive regulator of hedgehog signaling pathway along with the smoothened inhibitor patched protein (PTCH) and regulates vertebrate developmental processes, cellular proliferation and differentiation by activating transcription factor Gli. Smoothened mutants are known to cause sporadic basal cell carcinoma of the skin. It is known to express widely in heart and brain.Specifications
SMO | |
Polyclonal | |
Unconjugated | |
SMO | |
Gx, Protein Gx, SMO, SMOH, Smoothened homolog | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
6608 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.44 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q99835 | |
SMO | |
SMO Fusion Protein Ag14322 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction