Learn More
SMCO2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA553153
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: LKKKVIERLE DLCKNVELLS AKLRMYQMEA EDTDSHSSEE IDTEEMEALL PQAPASFLVQ KSPPRNTAWK R Highest antigen sequence identity to the following orthologs: Mouse - 41%, Rat - 38%.
SMCO2 is a protein coding gene.Specifications
SMCO2 | |
Polyclonal | |
Unconjugated | |
SMCO2 | |
C12orf70; single pass membrane protein with coiled-coil domains 2; single-pass membrane and coiled-coil domain-containing protein 2; single-pass membrane protein with coiled-coil domains 2; SMCO2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
341346 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
A6NFE2 | |
SMCO2 | |
Recombinant protein corresponding to Human SMCO2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only