Learn More
Fisherbrand™ Sliding Device
Use this accessory with phase contrast for inverted biological microscopes. Fisherbrand™ Sliding Devices are ergonomically designed for everyday use.
Brand: Fisherbrand™
Code : 46
Additional Details : Weight : 1.00000kg
Description
- Sliding Device
- Use with phase contrast
- For inverted biological microscope FB69196
Specifications
Sliding Device for Phase Contrast | |
inverted biological microscope FB69196 |