missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC4A3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA551351
Additional Details : Weight : 0.01000kg
Description
The antibody detects endogenous levels of total SLC4A3 protein.
Plasma membrane anion exchange protein of wide distribution. Mediates at least a part of the Cl-/HCO3- exchange in cardiac myocytes. Both BAE3 and CAE3 forms transport Cl-.Specifications
SLC4A3 | |
Polyclonal | |
Unconjugated | |
SLC4A3 | |
A930038D23Rik; AE 3; Ae3; Aep3; anion exchange protein 3; anion exchanger 3; anion exchanger 3 cardiac isoform; Anion exchanger 3, neuronal; B3rp3; B3RP-3; Band 3-related protein 3; CAE3/BAE3; Cardiac/brain band 3-like protein; neuronal band 3-like protein; SLC2C; Slc4a3; solute carrier family 4 (anion exchanger), member 3; solute carrier family 4 member 3; Solute carrier family 4, member 3, anion exchange protein 3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
20536, 24781, 6508 | |
-20°C | |
Liquid |
Western Blot | |
0.9 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
P16283, P23348, P48751 | |
SLC4A3 | |
Synthetic peptide of human SLC4A3 | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |