Learn More
SLC47A1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA554542
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: HANLKVNNVP RSGNSALPQD PLHPGCPENL EGILTNDVGK TGEPQSDQQM RQEEPLPEHP QDGAKLSRKQ Highest antigen sequence identity to the following orthologs: Mouse - 49%, Rat - 51%.
This gene is located within the Smith-Magenis syndrome region on chromosome 17. It encodes a protein of unknown function.Specifications
SLC47A1 | |
Polyclonal | |
Unconjugated | |
Slc47a1 | |
1300013J15Rik; AI036982; FLJ10847; H+/organic cation antiporter variant 1; H+/organic cation antiporter variant 2; hMATE-1; MATE1; MATE-1; MGC64822; mMATE1; mMATE-1; multidrug and toxin extrusion 1; multidrug and toxin extrusion protein 1; RGD1311123; rMATE-1; SLC47A1; solute carrier family 47 (multidrug and toxin extrusion), member 1; solute carrier family 47 member 1; solute carrier family 47, member 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55244 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96FL8 | |
Slc47a1 | |
Recombinant protein corresponding to Human SLC47A1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only