Learn More
SLC39A3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA559886
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: TFRKEKPSFI DLETFNAGSD VGSDSEYESP FMGGARGHAL YVEPHGHGPS LSVQGL Highest antigen sequence identity to the following orthologs: Mouse - 68%, Rat - 68%.
The zinc transporter ZIP3, also known as SLC39A3, is a member of a family of divalent ion transporters. Zinc is an essential ion for cells and plays significant roles in the growth, development, and differentiation. Similar to knock-outs of ZIP1 and ZIP2, ZIP3-null mice have no phenotypic differences compared to wild-type mice. Only when ZIP1, ZIP2, and ZIP3 genes are all eliminated and these mutant mice are fed a zinc-deficient diet do abnormalities such as reduced embryonic-membrane bound alkaline phosphatase activity and abnormal development occur, indicating that the ZIP1-3 proteins play an important, noncompensatory role when zinc is deficient. More recent studies have shown that ZIP2 and ZIP3 are down regulated in human prostate adenocarcinomatous glands, and may be important in the retention of zinc in the cellular compartment.Specifications
SLC39A3 | |
Polyclonal | |
Unconjugated | |
SLC39A3 | |
AI845814; SLC39A3; solute carrier family 39 (zinc transporter), member 3; solute carrier family 39 member 3; Zinc transporter ZIP3; Zip3; ZIP-3; Zrt- and Irt-like protein 3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
29985 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry, Western Blot | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9BRY0 | |
SLC39A3 | |
Recombinant protein corresponding to Human SLC39A3 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only