missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC35F2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA565636
Additional Details : Weight : 0.01000kg
Description
Immunogen sequence: MEADSPAGPGA PEPLAEGAAA EFSSLLRRIK GKLFTWN Highest antigen sequence identity to the following orthologs - mouse 55%, rat 55%.
SLC35F2 is a putative solute transporter.Specifications
SLC35F2 | |
Polyclonal | |
Unconjugated | |
SLC35F2 | |
1500009K05Rik; AU019213; HSNOV1; lung squamous cell cancer related protein LSCC-3; SLC35F2; Solute carrier family 35 member F2; solute carrier family 35, member F2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
54733 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Western Blot | |
0.10 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8IXU6 | |
SLC35F2 | |
Recombinant Human SLC35F2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
For Research Use Only