missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC25A14 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA566317
Additional Details : Weight : 0.01000kg
Description
Immunogen sequence: AAVIVSGHQKS TTVSHEMSGL NWKP Highest antigen sequence identity to the following orthologs - mouse 92%, rat 88%.
Mitochondrial uncoupling proteins (UCP) are members of the larger family of mitochondrial anion carrier proteins (MACP). UCPs separate oxidative phosphorylation from ATP synthesis with energy dissipated as heat, also referred to as the mitochondrial proton leak. UCPs facilitate the transfer of anions from the inner to the outer mitochondrial membrane and the return transfer of protons from the outer to the inner mitochondrial membrane. They also reduce the mitochondrial membrane potential in mammalian cells. Tissue specificity occurs for the different UCPs and the exact methods of how UCPs transfer H+/OH- are not known. UCPs contain the three homologous protein domains of MACPs. This gene is widely expressed in many tissues with the greatest abundance in brain and testis. The gene product has an N-terminal hydrophobic domain that is not present in other UCPs. Two splice variants have been found for this gene.Specifications
SLC25A14 | |
Polyclonal | |
Unconjugated | |
SLC25A14 | |
BMCP1; BMCP-1; brain mitochondrial carrier protein 1; MGC137654 protein; mitochondrial uncoupling protein 5; slc25a14; solute carrier family 25 (mitochondrial carrier, brain), member 14; solute carrier family 25 member 14; UCP 5; Ucp5; UCP5L; UCP5S; uncoupling protein 5; UNQ791/PRO1682; zgc:55596 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
9016 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O95258 | |
SLC25A14 | |
Recombinant Human SLC25A14 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
For Research Use Only