Learn More
SLC24A1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA558758
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: SSHQPIKLAS RDLSSEEMMM MSSSPSKPSS EMGGKMLVPQ ASVGSDEATL SMTVENIPSM PKRTAKMIPT TTKNNYSPTA AGTERRKED Highest antigen sequence identity to the following orthologs: Mouse - 51%, Rat - 28%.
SLC24A1 belongs to a family of potassium-dependent sodium/calcium exchangers. Members of this family have 2 large hydrophilic loops and 2 sets of multiple transmembrane-spanning segments (Schnetkamp, 2004 [PubMed 14770312]).Specifications
SLC24A1 | |
Polyclonal | |
Unconjugated | |
SLC24A1 | |
CSNB1D; HsT17412; KIAA0702; Na(+)/K(+)/Ca(2+)-exchange protein 1; NCKX; Nckx1; retinal Na/Ca,K exchanger; retinal rod Na+/Ca+/K+ exchanger; retinal rod Na-Ca+K exchanger; RODX; SLC24A1; sodium/calcium/potassium exchanger; sodium/potassium/calcium exchanger 1; solute carrier family 24 (sodium/potassium/calcium exchanger), member 1; solute carrier family 24 member 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
9187 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O60721 | |
SLC24A1 | |
Recombinant protein corresponding to Human SLC24A1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only