Learn More
SLC20A2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA555339
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: NSYTCYTAAI CGLPVHATFR AADSSAPEDS EKLVGDTVSY SKKRLRYDSY SSYCNAVAEA EIEAEEGGVE MKLASELADP DQPREDPAEE EKEEKDAPE Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 91%.
SLC20A2 Polyclonal antibody specifically detects SLC20A2 in Human samples. It is validated for Immunohistochemistry.Specifications
SLC20A2 | |
Polyclonal | |
Unconjugated | |
SLC20A2 | |
Ab1-188; fi23g11; Gibbon ape leukemia virus receptor 2; Glvr2; GLVR-2; hPit2; IBGC1; IBGC3; MLVAR; MolPit2; murine leukemia virus, amphotropic, receptor for; murine leukemia virus, amphotropic; receptor; phosphate transporter 2; PIT2; PIT-2; RAM1; RAM-1; receptor for amphitropic viruses 1; receptor for amphotropic viruses 1; slc20a2; Sodium-dependent phosphate transporter 2; solute carrier family 20 (phosphate transporter), member 2; solute carrier family 20 member 2; solute carrier family 20, member 2; type III sodium-dependent phosphate transporter; wu:fi23g11; zgc:152990 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
6575 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q08357 | |
SLC20A2 | |
Recombinant protein corresponding to Human SLC20A2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only