missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC14A1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA566162
Additional Details : Weight : 0.01000kg
Description
Immunogen sequence: MNGRSLIGGAG DARHGPVWKD PFGTKAGDAA RRGIARLSLA LADGSQEQE Highest antigen sequence identity to the following orthologs - mouse 60%, rat 32%.
SLC14A1 is a specialized low-affinity urea transporter. It mediates urea transport in erythrocytes.Specifications
SLC14A1 | |
Polyclonal | |
Unconjugated | |
SLC14A1 | |
2610507K20Rik; 3021401A05Rik; HsT1341; HUT11; JK; Kidd blood group; null; RACH1; RACH2; SLC14A1; SLC14A1 JK; solute carrier family 14 (urea transporter), member 1; solute carrier family 14 (urea transporter), member 1 (Kidd blood group); solute carrier family 14 member 1; solute carrier family 14 member 1 (Kidd blood group); urea transporter; urea transporter 1; Urea transporter B; urea transporter JK glycoprotein; urea transporter variant 2; Urea transporter, erythrocyte; urea transporter-B1; UrT2; UT1; UT-1; UT11; UT3; UT-B; Utb1; UT-B1; UTE | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
6563 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Western Blot | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q13336 | |
SLC14A1 | |
Recombinant Human SLC14A1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
For Research Use Only