Learn More
SLC10A7 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA559022
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: LSITPINEWL LKGLQTVGCM PPPVSSAVIL TKAVGGNEAA AIFNSAFGSF LVSKHSLTCL LQL Highest antigen sequence identity to the following orthologs: Mouse - 78%, Rat - 78%.
SLC10A7 (solute carrier family 10 (sodium/bile acid cotransporter family), member 7), also known as P7, is a 358 amino acid multi-pass membrane protein belonging to the sodium:bile acid symporter family. Existing as seven alternatively spliced isoforms, SLC10A7 is expressed at high levels in liver and lung, moderate levels in placenta, kidney, spleen, and thymus, and low levels in heart, prostate, and testis. A few members of the sodium:bile acid symporter family, such as NTCP (also known as SLC10A1) and Asbt (also known as SLC10A2), are involved in maintaining enterohepatic circulation of bile acids by mediating the first step of active bile transport through membrane barriers of liver and intestine. Other family members, including SLC10A6, play an important role in the cellular delivery of specific prohormones in testis, placenta, adrenal gland and other peripheral tissues. Orphan carriers such as SLC10A7 are uncharacterized and their functions unknown.Specifications
SLC10A7 | |
Polyclonal | |
Unconjugated | |
SLC10A7 | |
2410193C02Rik; C4orf13; Na(+)/bile acid cotransporter 7; P7; PSEC0051; SBF-domain containing protein; SLC10A7; Sodium/bile acid cotransporter 7; solute carrier family 10 (sodium/bile acid cotransporter family), member 7; solute carrier family 10 (sodium/bile acid cotransporter family), member 7-like; solute carrier family 10 member 7; solute carrier family 10, member 7 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
84068 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q0GE19 | |
SLC10A7 | |
Recombinant protein corresponding to Human SLC10A7 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only