missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SKIV2L Monoclonal Antibody (1E5), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA524551
Additional Details : Weight : 0.01000kg
Description
Peptide Sequence: DQLPNTLKQG IERVRAVAKR IGEVQVACGL NQTVEEFVGE LNFGLVEVVY EWARGMPFSE LAGLSGTPEG LVVRCIQRLA EMCRSLRGAA RLVGEPVLGA KMETAATLL
DEAD box proteins, characterized by the conserved motif Asp-Glu-Ala-Asp (DEAD), are putative RNA helicases. They are implicated in a number of cellular processes involving alteration of RNA secondary structure such as translation initiation, nuclear and mitochondrial splicing, and ribosome and spliceosome assembly. Based on their distribution patterns, some members of this family are believed to be involved in embryogenesis, spermatogenesis, and cellular growth and division. This gene encodes a DEAD box protein, which is a human homologue of yeast SKI2 and may be involved in antiviral activity by blocking translation of poly(A) deficient mRNAs. This gene is located in the class III region of the major histocompatibility complex.Specifications
SKIV2L | |
Monoclonal | |
Unconjugated | |
SKIV2L | |
170A, DDX13, HLP, SKI2, SKI2W, SKIV2, helicase SKI2W, helicase-like protein | |
Mouse | |
200 μL | |
Primary | |
Human | |
Antibody | |
IgG2a κ |
ELISA, Western Blot | |
1E5 | |
ascites with no preservative | |
Q15477 | |
SKIV2L | |
SKIV2L (NP_008860, 1125 a.a. ∽ 1233 a.a) partial recombit protein with GST tag. MW of the GST tag alone is 26 KDa. | |
RUO | |
6499 | |
Store at -20°C, Avoid Freeze/Thaw Cycles | |
Liquid |