Learn More
SIRPB2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA561637
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: GETLLLRCMV VGSCTDGMIK WVKVSTQDQQ EIYNFKRGSF PGVMPMIQRT SEPLNCDYSI YIHNVTREHT GTYHCVRFDG LSEHSEMKSD EGTSVLVKGA GDPEPDLWII QPQELVLGTT GDTVFLNCTV Highest antigen sequence identity to the following orthologs: Mouse - 36%, Rat - 39%.
The function of this protein remains unknown.Specifications
SIRPB2 | |
Polyclonal | |
Unconjugated | |
SIRPB2 | |
dJ776F14.2; Protein tyrosine phosphatase non-receptor type substrate 1-like 3; protein tyrosine phosphatase non-receptor type substrate protein; protein tyrosine phosphatase, non-receptor type substrate 1-like 3; PTPN1L; PTPNS1L3; signal regulatory protein beta 2; signal-regulatory protein beta 2; signal-regulatory protein beta-2; SIRPB2; SIRP-beta-2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
284759 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q5JXA9 | |
SIRPB2 | |
Recombinant protein corresponding to Human SIRPB2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only