missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NGF Recombinant Mouse Protein, Invitrogen™ Sino Biological™
This recombinant protein was expressed from a DNA sequence encoding the mature form of mouse NGF (NP_001106168.1), residues 122 to 241, with an initial Met at the N-terminus
£57.00 - £188.00
Specifications
For Use With (Application) | Protein Expression and Isolation, Enzyme and Protein Activity Assay, Protein Assay Control, Reference Standards and Accessories |
---|---|
Gene ID (Entrez) | 18049 |
Molecular Weight (g/mol) | 13.5kDa |
Storage Requirements | Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed. |
Structural Form | Native |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
13476286
|
Invitrogen
50385MNAC5 |
5μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
13456286
|
Invitrogen
50385MNAC25 |
5 x 5μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
13486286
|
Invitrogen
50385MNAC50 |
50μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Nerve growth factor (NGF), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in performing cell based assays. It can also be used as a protein standard, or in other research applications.- Activity: Measured in a cell proliferation assay using TF1 human erythroleukemic cells; the ED50 for this effect is 0.2-2.0ng/mL
- Formulation: Lyophilized in 50mM Na acetate buffer, pH 5.5, 150mM NaCl, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
- Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible
Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.
Specifications
Protein Expression and Isolation, Enzyme and Protein Activity Assay, Protein Assay Control, Reference Standards and Accessories | |
13.5kDa | |
Native | |
Ngf, Ngfb | |
NGF | |
Mammalian | |
Lyophilized | |
≥95% by SDS-PAGE |
18049 | |
Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed. | |
<1 EU/μg | |
Nerve growth factor | |
Untagged | |
Growth Factors & Receptors | |
Neurotrophic Factors |
For Research Use Only. Not for use in diagnostic procedures.