missing translation for 'onlineSavingsMsg'
Learn More

Silver Nitrate, Honeywell Fluka™

Product Code. 15660950
Click to view available options
Quantity:
50 g
100 g
500 g
1 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

Meets analytical specification of BP, Ph. Eur., USP, 99.8-100.5%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7761-88-8
Molecular Formula AgNO3
Molecular Weight (g/mol) 169.87
MDL Number MFCD00003414
InChI Key SQGYOTSLMSWVJD-UHFFFAOYSA-N
Synonym silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol
PubChem CID 24470
ChEBI CHEBI:32130
IUPAC Name silver(1+) nitrate
SMILES [Ag+].[O-][N+]([O-])=O

Specifications

Quantity 1 kg
Linear Formula AgNO3
UN Number 1493
Solubility Information Soluble in ethanol, 20.8g/L, Soluble in benzene, 2.2g/L, Soluble in methanol, 35g/L
Formula Weight 169.87g/mol
Percent Purity 99.8 to 100.5%
Density Vapor Density: 5.8 (vs air)
Chemical Name or Material Silver Nitrate, Meets analytical specification of Ph. Eur. BP, USP
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'