missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Silver(I) Oxide, Honeywell™
Click to view available options
Quantity:
10 g
50 g
250 g
Description
- ≥95% for general laboratory use.
Specifications
Specifications
Quantity | 250 g |
UN Number | 1479 |
Formula Weight | 231.74g/mol |
Percent Purity | 99% |
Grade | ReagentPlus™ |
Chemical Name or Material | Silver(I) Oxide |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'