missing translation for 'onlineSavingsMsg'
Learn More
Silicon carbide powder, coarse, 46 grit, Thermo Scientific Chemicals Product Code.: 11411798

Silicon carbide powder, coarse, 46 grit, Thermo Scientific Chemicals

Product Code. 11411798
10,000 g
Click to view available options
Quantity:
10,000 g
2500 g
500 g
This item is not returnable. View return policy

Product Code. 11411798

Brand: Thermo Scientific Alfa Aesar A14470.0C

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Silicon carbide is used in abrasives, in polishing and grinding. It is widely used in applications calling for high endurance, such as automobile brakes, car clutches and ceramic plates in bulletproof vests. Electronic applications of silicon carbide are as light emitting diodes and sensors. It is utilized for the production of fertile, transgenic maize plants. It compositely plays an important role in the crack-healing behavior.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Silicon carbide is used in abrasives, in polishing and grinding. It is widely used in applications calling for high endurance, such as automobile brakes, car clutches and ceramic plates in bulletproof vests. Electronic applications of silicon carbide are as light emitting diodes and sensors. It is utilized for the production of fertile, transgenic maize plants. It compositely plays an important role in the crack-healing behavior.

Solubility
Soluble in molten sodium hydroxide, potassium hydroxide and in molten iron.

Notes
Keep it away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 409-21-2
Molecular Formula CSi
Molecular Weight (g/mol) 40.10
MDL Number MFCD00049531
InChI Key HBMJWWWQQXIZIP-UHFFFAOYSA-N
Synonym silicon carbide, carborundum, silicon monocarbide, carborundeum, tokawhisker, betarundum, carbolon, nicalon, silundum, carbon silicide
PubChem CID 9863
ChEBI CHEBI:29390
SMILES [C-]#[Si+]

Specifications

Quantity 10,000 g
Solubility Information Soluble in molten sodium hydroxide,potassium hydroxide and in molten iron.
IUPAC Name methanidylidynesilylium
Formula Weight 40.1
Odor Odorless
Density 3.16
Chemical Name or Material Silicon carbide powder, coarse, 46 grit

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Silicon carbide powder, coarse, 46 grit, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.