Learn More
BINDER™ Right Access Port (10 to 30 mm)
For Use With: BINDER units ED 23, ED 400
Brand:  BINDER™ 8012-0536
Product Code. 15671776
  • £515.00 / Each
Estimated Shipment: 18-07-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • For use on right
  • Includes silicone plug
  • For use with a wide range of BINDER™ incubators and chambers
Specifications
Specifications
Access port with silicone plug - 30 mm, right
Steel
Right
30 mm
BINDER Chambers ED 23, ED 400
Access Port
Silicone plug
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
BINDER™ Right Access Port (10 to 30 mm) > Diameter: 30 mm, For Use With: ED 23 and ED 400