missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Memmert™ Shelf Reversible For 2 Heights (30 Or 60 mm), For One45
Use these shelves with Memmert water baths. Memmert™ Reversible Shelves for Water Baths are available in a wide range of sizes to fit your needs.
Brand: Memmert™ ONE45E02896
This item is not returnable.
View return policy
Description
- Reversible Shelves
- For Memmert Water Baths
Specifications
29 L and 45 L Baths | |
Reversible Shelf |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Memmert™ Shelf Reversible For 2 Heights (30 Or 60 mm), For One45 >
Spot an opportunity for improvement?Share a Content Correction