Learn More
Sharpsguard™ anatomical 2.5 Waste Container
For the safe disposal of anatomical and placenta waste. Sharpsguard™ anatomical 2.5 Waste Containers each come with a placenta bag.
Brand: Daniels Healthcare DD912
Code : 02
Additional Details : Weight : 9.63000kg
Description
- Rigid polypropylene construction and wall thickness
- Offers outstanding protection
- When container is full to the fill line it must be locked and disposed of safely
- All have clear British Standard markings, user instructions and a visible fill line
Specifications
Anatomical Waste | |
2.21 L | |
Red, Yellow |