Learn More
BD Vacutainer Sharps Container
For point-of-use disposal to ensure maximum safety.
Brand: BD Vacutainer 367202
Code : NEW
Additional Details : Weight : 0.87000kg
Description
- Available in a range of sizes and shapes.
- Designed for point-of-use disposal to ensure maximum safety.
- Provides an environment and an approach that is focused on safety to help reduce the risks of blood exposure and needle stick injuries associated with the disposal process.
- Compatible brackets for placement on flat surfaces, secured to walls, or for placement on medication carts.
Specifications
0.7 L | |
89 x 74 x 168 mm | |
50/Pk. |