Learn More
Thermo Scientific™ Shandon™ Reader barcode, with UK plug
Brand:  Thermo Scientific™ A78210103
Product Code. 12668146
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
Included with unit: CD-ROM with unlicenced demo copy of Thermo Scientific Shandon MicroWriter™ software; Dispenser hoppers – six included; Pre-filled cassette tube sample pack; MicroWriter™ specific RS232 cable; Power cord; Operator manual.
Uses foil tape and a heated stylus to imprint characters that maintain legibility even through processing solutions
  • Made to withstand the workload of busy laboratories, marking approximately 20,000 characters per roll of tape
  • Printing can be inverted for reading in either direction
  • Change cassette colour and style easily with fully automated dispensing and labelling process
  • Easy to change hoppers provide the flexibility of allowing an unlimited number of colours and cassettes
  • Imprints, letters, numbers, Roman numerals and special characters on the cassettes in four font sizes
  • Ability to print barcodes on cassettes
  • Prints on all 45° flat-faced cassettes
  • Choice of collection systems makes organising cassettes a seamless operation in any laboratory environment
Carousel-type cassette marking system that permanently marks tissue cassettes with easy to read black type.
TRUSTED_SUSTAINABILITY
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Thermo Scientific™ Shandon™ Reader barcode, with UK plug >