missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SH3GL2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
Antigen | SH3GL2 |
---|---|
Concentration | 0.28 mg/mL |
Applications | Immunoprecipitation, Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16810154
|
Proteintech
12435-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16800154
|
Proteintech
12435-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
SH3GL2 | |
Immunoprecipitation, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
Mouse, Human, Rat | |
O35179, Q62420, Q99962 | |
116743, 20404, 6456 | |
SH3GL2 Fusion Protein Ag3114 | |
Primary | |
-20°C | |
SH3GL2 |
0.28 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
CNSA2, EEN B1, Endophilin 1, Endophilin A1, SH3 domain GRB2 like 2, SH3 domain protein 2A, SH3D2A, SH3GL2, SH3P4 | |
SH3GL2 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
SH3GL2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech