missing translation for 'onlineSavingsMsg'
Learn More

SGTB Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

£135.00 - £349.00

Specifications

Antigen SGTB
Concentration 133 μg/mL
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17541336
View Documents
Proteintech
15895-1-AP-20UL
20 μL
£135.00
20µL
Please to purchase this item. Need a web account? Register with us today!
17531336
View Documents
Proteintech
15895-1-AP-150UL
150 μL
£349.00
150µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The SGTB antibody from Proteintech is a rabbit polyclonal antibody to a recombinant protein of human SGTB. This antibody recognizes human, mouse, rat antigen. The SGTB antibody has been validated for the following applications: WB, ELISA analysis.

Specifications

Specifications

SGTB
Western Blot
Unconjugated
Human, Mouse, Rat
IgG
133 μg/mL
Polyclonal
Rabbit
54557
Primary
Videos
SDS
Documents

Documents

Product Certifications