missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human SGSM1 Synthetic Peptide
Synthetic Peptide
Brand: Invitrogen™ PEP1480
Code : NEW
Additional Details : Weight : 0.01000kg
Description
SGSM1 Synthetic Peptide, PEP-1480, from Invitrogen.
This peptide corresponds to 17 amino acid peptide near the center of human SGSM1.PEP-1480 can be used as a blocking peptide with polyclonal antibody PA5-34438.Specifications
129049 | |
purified | |
Human SGSM1 | |
-20°C | |
Liquid | |
Blocking Assay, Control | |
No | |
Peptide | |
RUO - research use only |
Human | |
Q2NKQ1 | |
200 μg/mL | |
Synthetic Peptide | |
PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2 | |
SGSM1 | |
0.02% sodium azide | |
50 μg | |
PBS |