missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SGE™ Replacement Needles for 0.5 - 1μL Syringes
For use with 0.5 to 1μL syringes
Brand: SGE™ 033772
Code : NEW
Additional Details : Weight : 0.01000kg
Description
For removable-needle syringes or additional replacements, contact your Fisher Scientific Customer Service Representative.
Specifications
Stainless Steel | |
0.006 in. | |
0.15 mm | |
2 in. | |
Cone |
23 G | |
0.02 in. | |
0.5 mm | |
Replacement Needle | |
Syringe Needles (0.5μL), Syringes |