missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Sertraline hydrochloride
Click to view available options
Quantity:
1 g
5 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 245°C |
Quantity | 1 g |
Merck Index | 15,8606 |
Solubility Information | Solubility in water: .. Other solubilities: soluble in methanol |
Formula Weight | 342.69 |
Chemical Name or Material | Sertraline hydrochloride |
Safety and Handling
Signal Word
- Warning
- Acute toxicity Category 4
- ACUTE AQUATIC Acute 1
- H302-Harmful if swallowed.
- H400-Very toxic to aquatic life.
- P264-Wash thoroughly after handling.
- P273-Avoid release to the environment.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P391-Collect spillage.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'