missing translation for 'onlineSavingsMsg'
Learn More

Semperit™ Semperguard™ Vinyl Powdered Disposable Gloves

Product Code. 15503526
Click to view available options
Size:
Small
Medium
Large
X-Large

Semperguard™ Vinyl Powdered Disposable Gloves are safe for use with fat-free foodstuffs.

  • Pre-powdered
  • Vinyl construction
  • Free from latex proteins and accelerants
  • Free from DOP
  • Suitable for fat-free food contact
  • CE Category I minimal risks
  • Transparent in color

Specifications

Certifications/Compliance ISO 9001, ISO 13485
Classification CE Category I
Cut Style Rolled
Color Transparent
Disposable Single-use
Latex Free Yes
Material Vinyl
Powdered Powdered
Product Type Disposable Gloves
Size X-Large
Thickness (Metric) 0.15 mm
For Use With (Application) Personal Protection, Professional Cleaning/Hygiene, Alternate Care
Glove Cuff Style Rolled
Packaging Type Dispenser Box
Quantity 100 Pk.
Length (Metric) 240 mm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.