Learn More
SELK Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 12561-1-AP-150UL
 View more versions of this product
Product Code. 16800334
  • £349.00 / 150µL
Explore more special offers
Description
Description
SELK is a selenoprotein, containing a selenocysteine residue at its active site. SELK is localized to the endoplasmic reticulum and is highly expressed in the heart, where it may function as an antioxidant. SELK can bind to proteins such as the ER-associated degradation (ERAD) components p97 ATPase, Derlin-1 and -2, and SelS as well as proteins in the oligosaccharyltransferase complex, suggesting that SELK may be involved in the Derlin-dependent ERAD of glycosylated misfolded proteins. SELK is also thought to be important in Ca2+ flux in immune cells, and is cleaved by m-calpain in resting macrophages. When these macrophages are activated through different Toll-like receptors, this cleavage is inhibited
Specifications
Specifications
SELK
Polyclonal
Unconjugated
SELENOK
HSPC030, HSPC297, selenoprotein K, SELK
Rabbit
Antigen Affinity Chromatography
RUO
290549, 58515, 80795
-20°C
Liquid
Western Blot
0.15 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P59798, Q9JLJ1, Q9Y6D0
Selenok
SELK Fusion Protein Ag3143
150 μL
Primary
Human, Mouse, Rat