Learn More
DWK Life Sciences Sealing Ring, for polished spherical joints (VITON)
For sealing ground joints without ground joints DURAN™, especially for vacuum applications.
Brand: DWK Life Sciences 1155356
Additional Details : Weight : 1.00000kg
Description
Ball joints without ground joints only seal via the sealing ring. VITON sealing rings are used in vacuum applications.
- Due to the material properties, it can be used for work with high demands on the chemical resistance of the sealing ring.
- Sealing ring made of VITON
Specifications
Sealing of non-sanded spherical ground joints | |
10/Pk. | |
Viton | |
19 NS |