missing translation for 'onlineSavingsMsg'
Learn More
Saponin, From Saponegin Glycosides (Off-White Powder/Practical Grade), MP Biomedicals Product Code.: 11496222

Saponin, From Saponegin Glycosides (Off-White Powder/Practical Grade), MP Biomedicals

Product Code. 11496222 Shop All MP Biomedicals Products
25 g
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy

Product Code. 11496222

Brand: MP Biomedicals 0210285525

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Plant origin, Saponegin glycosides

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 637-39-8
Molecular Formula C6H16ClNO3
Molecular Weight (g/mol) 185.648
InChI Key HHLJUSLZGFYWKW-UHFFFAOYSA-N
Synonym triethanolamine hydrochloride, tea-hydrochloride, triethanolammonium chloride, 2,2',2-nitrilotriethanol hydrochloride, ethanol, 2,2',2-nitrilotris-, hydrochloride, 2,2',2-nitrilotrisethanol hydrochloride, unii-r297uj9qdy, tris 2-hydroxyethyl ammonium chloride, tris 2-hydroxyethyl amine hydrochloride, r297uj9qdy
PubChem CID 101814
IUPAC Name 2-[bis(2-hydroxyethyl)amino]ethanol;hydrochloride
SMILES C(CO)N(CCO)CCO.Cl

Specifications

Color White
Quantity 25 g
Grade Practical
Physical Form Powder
Chemical Name or Material Saponin, Cell biology
compliance-icons
Product Identifier
  • Saponin, Practical Grade
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P405-Store locked up.
  • P501-Dispose of contents/container to in accordance with local/regional/national/international regulations..
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Saponin, From Saponegin Glycosides (Off-White Powder/Practical Grade), MP Biomedicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.