Learn More
Salicylaldehyde azine, 97%, Thermo Scientific Chemicals
£65.43 - £257.28
Chemical Identifiers
CAS | 959-36-4 |
---|---|
Molecular Formula | C14H12N2O2 |
Molecular Weight (g/mol) | 240.262 |
MDL Number | MFCD00043496 |
InChI Key | SPEXYYIULCBQJR-UHFFFAOYSA-N |
Synonym | salicylaldehyde azine, salicylalazine, salicylaldazine, benzaldehyde, 2-hydroxy-, 2-hydroxyphenyl methylene hydrazone, 2-hydroxybenzaldehyde azine, 2-1e-e-2-2-hydroxyphenyl methylidene hydrazin-1-ylidene methyl phenol, 2,2'-1,2-hydrazinediylidenedi methylylidene diphenol, benzaldehyde, 2-hydroxy-, 2-2-hydroxyphenyl methylene hydrazone, 1,2-bis e-salicylidene hydrazine, 6-6-oxo-1-cyclohexa-2,4-dienylidene methylhydrazo methylidene-1-cyclohexa-2,4-dienone |
PubChem CID | 6849893 |
IUPAC Name | 6-[[2-[(6-oxocyclohexa-2,4-dien-1-ylidene)methyl]hydrazinyl]methylidene]cyclohexa-2,4-dien-1-one |
SMILES | C1=CC(=CNNC=C2C=CC=CC2=O)C(=O)C=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11498793
|
Thermo Scientific Alfa Aesar
A10527.09 |
10 g |
£65.43
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11408803
|
Thermo Scientific Alfa Aesar
A10527.18 |
50 g |
£257.28
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Salicylaldehyde azine is used in the preparation of 2-oxo-2H-1-benzopyran-3-carboxylic acid 2-(2-oxo-2H-1-benzopyran-3-carbonyl)hydrazide by reacting with propadienedione . It has anthelmintic activity.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsSalicylaldehyde azine is used in the preparation of 2-oxo-2H-1-benzopyran-3-carboxylic acid 2-(2-oxo-2H-1-benzopyran-3-carbonyl)hydrazide by reacting with propadienedione . It has anthelmintic activity.
Solubility
Soluble in dimethyl sulfoxide, methanol and ethyl acetate. Slightly soluble in water.
Notes
Incompatible with strong oxidizing agents.
Chemical Identifiers
959-36-4 | |
240.262 | |
SPEXYYIULCBQJR-UHFFFAOYSA-N | |
6849893 | |
C1=CC(=CNNC=C2C=CC=CC2=O)C(=O)C=C1 |
C14H12N2O2 | |
MFCD00043496 | |
salicylaldehyde azine, salicylalazine, salicylaldazine, benzaldehyde, 2-hydroxy-, 2-hydroxyphenyl methylene hydrazone, 2-hydroxybenzaldehyde azine, 2-1e-e-2-2-hydroxyphenyl methylidene hydrazin-1-ylidene methyl phenol, 2,2'-1,2-hydrazinediylidenedi methylylidene diphenol, benzaldehyde, 2-hydroxy-, 2-2-hydroxyphenyl methylene hydrazone, 1,2-bis e-salicylidene hydrazine, 6-6-oxo-1-cyclohexa-2,4-dienylidene methylhydrazo methylidene-1-cyclohexa-2,4-dienone | |
6-[[2-[(6-oxocyclohexa-2,4-dien-1-ylidene)methyl]hydrazinyl]methylidene]cyclohexa-2,4-dien-1-one |
Specifications
959-36-4 | |
97% | |
MFCD00043496 | |
salicylaldehyde azine, salicylalazine, salicylaldazine, benzaldehyde, 2-hydroxy-, 2-hydroxyphenyl methylene hydrazone, 2-hydroxybenzaldehyde azine, 2-1e-e-2-2-hydroxyphenyl methylidene hydrazin-1-ylidene methyl phenol, 2,2'-1,2-hydrazinediylidenedi methylylidene diphenol, benzaldehyde, 2-hydroxy-, 2-2-hydroxyphenyl methylene hydrazone, 1,2-bis e-salicylidene hydrazine, 6-6-oxo-1-cyclohexa-2,4-dienylidene methylhydrazo methylidene-1-cyclohexa-2,4-dienone | |
SPEXYYIULCBQJR-UHFFFAOYSA-N | |
6-[[2-[(6-oxocyclohexa-2,4-dien-1-ylidene)methyl]hydrazinyl]methylidene]cyclohexa-2,4-dien-1-one | |
6849893 | |
97% |
216°C to 222°C | |
C14H12N2O2 | |
1913597 | |
Soluble in dimethyl sulfoxide,methanol and ethyl acetate. Slightly soluble in water. | |
C1=CC(=CNNC=C2C=CC=CC2=O)C(=O)C=C1 | |
240.262 | |
240.26 | |
Salicylaldehyde Azine |
Safety and Handling
GHS H Statement
H315-H319-H335
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
missing translation for 'einecsNumber' : 213-500-0
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Salicylaldehyde azine
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
Precautionary Statement
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.