missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ SAHARA S21SS Stainless Steel Heated Bath

Durable, seamless stainless steel bath for high temperature applications

Brand:  Thermo Scientific™ 1701210

Product Code. 11690931

  • £1067.96 / Each
Estimated Shipment: 06-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Bath volume: 7-17L
  • Mateial: Stainless steel
  • Ambient +13 to 200°C


TRUSTED_SUSTAINABILITY
Specifications

Specifications

21 L
Stainless Steel
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers