Learn More
Fisherbrand™ Safety Coated Clear Boston Round Amber Glass Bottles with White Polypropylene Caps
Type III soda-lime glass is ideal for solvent, chemical or sample storage with plastisol coating for safety are easier to grip and harder to break.
£74.75 - £155.00
Specifications
Color Amber
Material Glass (Type 3)
Closure Color White
Closure Included Yes
Mouth Narrow
View More Specs
Products 4
Product Code Brand Capacity (English) Capacity (Metric) Closure Liner Price Quantity & Availability  
Product Code Brand Capacity (English) Capacity (Metric) Closure Liner Price Quantity & Availability  
16154497
View accessory products View Documents
Fisherbrand™
FB02911894
4 oz. 120 mL PTFE/Polyethylene Foam
£103.00
Pack of 24
Estimated Shipment: 22-05-2025
to see stock.
Add to basket
16114467
View accessory products View Documents
Fisherbrand™
FB02911815
8 oz. 240 mL PTFE/Polyethylene Foam
£74.75
Pack of 12
Estimated Shipment: 09-01-2025
to see stock.
Add to basket
16124467
View accessory products View Documents
Fisherbrand™
FB02911817
16 oz. 480 mL PTFE/Polyethylene Foam
£83.85
Pack of 12
Estimated Shipment: 31-01-2025
to see stock.
Add to basket
16104467
View accessory products View Documents
Fisherbrand™
FB02911814
32 oz. 950 mL PTFE/Polyethylene Foam
£155.00
Pack of 12
Estimated Shipment: 18-02-2025
to see stock.
Add to basket
Description
Description
  • Coated in layer of plastisol
  • Shatter-resistant coating reduces chance of breakage
  • If broken, safety coating will contain contents long enough for disposal
  • Non-slip coating also allows greater ease of stability, wet or dry
  • Not autoclavable
Specifications
Specifications
Amber
White
Narrow
PTFE/Polyethylene Foam
Glass (Type 3)
Yes
Safety-coated Boston Round Bottle
Polypropylene