missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-Tetrahydro-2-furoic acid, 98+%, ee 98%, Thermo Scientific™

Product Code. 11308716
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 87392-07-2
Molecular Formula C5H8O3
Molecular Weight (g/mol) 116.116
MDL Number MFCD00272639
InChI Key UJJLJRQIPMGXEZ-BYPYZUCNSA-N
Synonym s-tetrahydrofuran-2-carboxylic acid, s---2-tetrahydrofuroic acid, s---tetrahydro-2-furoic acid, 2s-oxolane-2-carboxylic acid, s-tetrahydro-2-furoic acid, s---tetrahydrofuran-2-carboxylic acid, unii-0i2b46k2sj, tetrahydro-2-furoic acid,-, 2-furancarboxylic acid, tetrahydro-, 2s, s---2-carboxytetrahydrofuroic acid
PubChem CID 446666
ChEBI CHEBI:45960
IUPAC Name (2S)-oxolane-2-carboxylic acid
SMILES C1CC(OC1)C(=O)O

Specifications

Density 1.213
Boiling Point 244°C
Quantity 25 g
Refractive Index 1.46
UN Number UN3265
Beilstein 4658738
Formula Weight 116.12
Percent Purity ≥98%
Assay ee 98%
Chemical Name or Material (S)-(-)-Tetrahydro-2-furoic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.