missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-3-Chloro-1-phenyl-1-propanol, 98+%, Thermo Scientific Chemicals

Product Code. 15431018
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100306-34-1
Molecular Formula C9H11ClO
Molecular Weight (g/mol) 170.636
MDL Number MFCD00013309
InChI Key JZFUHAGLMZWKTF-VIFPVBQESA-N
Synonym s---3-chloro-1-phenyl-1-propanol, s-3-chloro-1-phenylpropan-1-ol, s-3-chloro-1-phenyl-1-propanol, 1s-3-chloro-1-phenylpropan-1-ol, 1s-3-chloro-1-phenyl-propan-1-ol, s-3-chloro-1-phenylpropanol, s-3-chloro-1-phenyl-propan-1-ol, pubchem6275
PubChem CID 2777894
IUPAC Name (1S)-3-chloro-1-phenylpropan-1-ol
SMILES C1=CC=C(C=C1)C(CCCl)O

Specifications

Melting Point 58°C to 60°C
Quantity 1 g
Optical Rotation −26° (c=1 in Chloroform)
Formula Weight 170.64
Percent Purity ≥98%
Chemical Name or Material (S)-(-)-3-Chloro-1-phenyl-1-propanol

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'